4-(tert-Butylsulfanyl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044643-500MG 044643-1G 044643-5G
101866-870EA 89.23 USD
101866-870 101866-872 101866-874
4-(tert-Butylsulfanyl)benzenecarbaldehyde ≥95%
4-(tert-Butylsulfanyl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄OS
MW: 194.3 g/mol
MDL Number: MFCD08443944
CAS Number: 88357-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR