1-Bromo-4-(tert-butylsulfanyl)benzene ≥95%

Supplier: Matrix Scientific

044638-500MG 044638-1G 044638-5G
101866-840EA 78.46 USD
101866-840 101866-842 101866-844
1-Bromo-4-(tert-butylsulfanyl)benzene ≥95%
1-Bromo-4-(tert-butylsulfanyl)benzene

Hazard: Irritant

Formula: C₁₀H₁₃BrS
MW: 245.19 g/mol
MDL Number: MFCD04973729
CAS Number: 25752-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR