Ethyl-1-(5-formyl-2-pyridinyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044633-500MG 044633-1G 044633-5G
101866-822EA 556.92 USD
101866-822 101866-824 101866-826
Ethyl-1-(5-formyl-2-pyridinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(5-formyl-2-pyridinyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃
MW: 262.31 g/mol
MDL Number: MFCD04973727
CAS Number: 886360-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR