6-(2,6-Dimethylmorpholino)nicotinaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
6-(2,6-Dimethylmorpholin-4-yl)pyridine-3-carbaldehyde
044632-500MG
044632-1G
044632-5G
101866-816EA
384.62
USD
101866-816
101866-818
101866-820
6-(2,6-Dimethylmorpholino)nicotinaldehyde ≥95%
6-(2,6-Dimethylmorpholino)nicotinaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ MW: 220.27 g/mol |
MDL Number:
MFCD04973726 CAS Number: 886360-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...