tert-Butyl-8-benzyl-2,8-diazaspiro[4.5]decane-2-carboxylate ≥95%

Supplier: Matrix Scientific

044570-500MG 044570-1G
101866-504EA 1109.23 USD
101866-504 101866-506
tert-Butyl-8-benzyl-2,8-diazaspiro[4.5]decane-2-carboxylate ≥95%
tert-Butyl-8-benzyl-2,8-diazaspiro[4.5]decane-2-carboxylate

Hazard: Irritant

Formula: C₂₀H₃₀N₂O₂
MDL Number: MFCD12400945
CAS Number: 336191-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR