8-Benzyl-2,8-diazaspiro[4.5]decane ≥95%

Supplier: Matrix Scientific

044569-500MG 044569-1G
101866-500EA 538.46 USD
101866-500 101866-502
8-Benzyl-2,8-diazaspiro[4.5]decane ≥95%
8-Benzyl-2,8-diazaspiro[4.5]decane

Hazard: Irritant

Formula: C₁₅H₂₂N₂
MDL Number: MFCD09910205
CAS Number: 336191-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR