[3-(tert-Butoxy)-2-(methoxyimino)-3-oxopropyl](triphenyl)phosphonium bromide ≥95%

Supplier: Matrix Scientific

044542-500MG 044542-1G
101866-390EA 1109.23 USD
101866-390 101866-392
[3-(tert-Butoxy)-2-(methoxyimino)-3-oxopropyl](triphenyl)phosphonium bromide ≥95%
[3-(tert-Butoxy)-2-(methoxyimino)-3-oxopropyl](triphenyl)phosphonium bromide

Hazard: Irritant

Formula: C₂₆H₂₉BrNO₃P
MDL Number: MFCD11553019
CAS Number: 1231928-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR