Ethyl-2-(2,4-dichloro-5-nitrophenoxy)acetate ≥95%

Supplier: Matrix Scientific

044497-500MG 044497-1G 044497-5G
101866-176EA 278.46 USD
101866-176 101866-178 101866-180
Ethyl-2-(2,4-dichloro-5-nitrophenoxy)acetate ≥95%
Ethyl-2-(2,4-dichloro-5-nitrophenoxy)acetate

Hazard: Irritant

Formula: C₁₀H₉Cl₂NO₅
MDL Number: MFCD09972207
CAS Number: 501008-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR