2,4-Dichlorophenyl ethyl carbonate ≥95%

Supplier: Matrix Scientific

044494-500MG 044494-1G 044494-5G
101866-158EA 181.54 USD
101866-158 101866-160 101866-162
2,4-Dichlorophenyl ethyl carbonate ≥95%
2,4-Dichlorophenyl ethyl carbonate

Hazard: Irritant

Formula: C₉H₈Cl₂O₃
MDL Number: MFCD08056625
CAS Number: 22876-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR