2-Fluoro-6-pyridone ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-6-hydroxypyridine, 6-Fluoropyridin-2-ol
041024-1G
041024-5G
101850-634EA
16.92
USD
101850-634
101850-636
2-Fluoro-6-pyridone ≥98%
2-Fluoro-6-pyridone
Hazard: Irritant
Formula:
C₅H₄FNO MW: 113.09 g/mol |
MDL Number:
MFCD03092938 CAS Number: 50543-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...