3,5-Dimethylisoxazole-4-carbonyl chloride

Supplier: Matrix Scientific

040896-1G 040896-5G 040896-25G
101850-382EA 36.92 USD
101850-382 101850-384 101850-386
3,5-Dimethylisoxazole-4-carbonyl chloride
3,5-Dimethylisoxazole-4-carbonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₆ClNO₂
MW: 159.57 g/mol
MDL Number: MFCD00052555
CAS Number: 31301-45-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR