Ethyl-5-methylisoxazole-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 5-methylisoxazole-3-carboxylate

040764-1G 040764-5G
101850-130EA 49.23 USD
101850-130 101850-132
Ethyl-5-methylisoxazole-3-carboxylate
Ethyl 5-methylisoxazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD00085080
CAS Number: 3209-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR