N-(tert-Butyl)-2-chloroacetamide

Supplier: Matrix Scientific

040455-1G 040455-5G
101849-544EA 170.77 USD
101849-544 101849-546
N-(tert-Butyl)-2-chloroacetamide
N-(tert-Butyl)-2-chloroacetamide

Hazard: Irritant

Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
MDL Number: MFCD00032565
CAS Number: 15678-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR