2-(1,3-Thiazol-2-yl)ethanamine dihydrochloride

Supplier: Matrix Scientific

037001-500MG 037001-1G
101842-486EA 92.31 USD
101842-486 101842-488
2-(1,3-Thiazol-2-yl)ethanamine dihydrochloride
2-(1,3-Thiazol-2-yl)ethanamine dihydrochloride

Hazard: Irritant

Formula: C₅H₁₀Cl₂N₂S
MDL Number: MFCD13250086
CAS Number: 56933-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR