Methyl-5-(chloromethyl)-2-furoate

Supplier: Matrix Scientific

Synonyms: Methyl 5-(chloromethyl)-2-furancarboxylate, Methyl 5-(chloromethyl)furan-2-carboxylate

036501-5G 036501-25G
101841-500EA 80 USD
101841-500 101841-502
Methyl-5-(chloromethyl)-2-furoate
Methyl-5-(chloromethyl)-2-furoate

Hazard: Corrosive/Stored under Argon/Moisture Sensitive/Refrigerate

Formula: C₇H₇ClO₃
MW: 174.58 g/mol
Boiling Pt: 32-36°C
MDL Number: MFCD00020922
CAS Number: 2144-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR