3,5-Dimethyl-4-nitropyrazole ≥95%
Supplier: Matrix Scientific
Synonyms:
3,5-Dimethyl-4-nitro-1H-pyrazole
036453-1G
036453-5G
101841-398EA
15.38
USD
101841-398
102551-186
3,5-Dimethyl-4-nitropyrazole ≥95%
3,5-Dimethyl-4-nitropyrazole
Hazard: Irritant
Formula:
C₅H₇N₃O₂ MW: 141.13 g/mol |
MDL Number:
MFCD00052513 CAS Number: 14531-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...