4-(Chloromethyl)-3,5-dimethylisoxazole ≥95%

Supplier: Matrix Scientific

036451-5G 036451-25G
101841-392EA 46.15 USD
101841-392 101841-394
4-(Chloromethyl)-3,5-dimethylisoxazole ≥95%
4-(Chloromethyl)-3,5-dimethylisoxazole

Hazard: Irritant

Formula: C₆H₈ClNO
MW: 145.59 g/mol
MDL Number: MFCD00003154
CAS Number: 19788-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR