4-Methoxy-3-[(4-methyl-1H-pyrazol-1-yl)methyl]-benzoic acid

Supplier: Matrix Scientific
Warning

036421-500MG
101841-324EA 387.69 USD
101841-324
4-Methoxy-3-[(4-methyl-1H-pyrazol-1-yl)methyl]-benzoic acid
4-Methoxy-3-[(4-methyl-1H-pyrazol-1-yl)methyl]-benzoic acid
Formula: C₁₃H₁₄N₂O₃
MW: 246.27 g/mol
MDL Number: MFCD08277095
CAS Number: 956208-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR