3-Cyano-4-fluorobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-formylbenzonitrile

036290-5G 036290-25G
101841-064EA 35.38 USD
101841-064 101841-066
3-Cyano-4-fluorobenzaldehyde ≥95%
3-Cyano-4-fluorobenzaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₈H₄FNO
MW: 149.1219 g/mol
Melting Pt: 80…84 °C
MDL Number: MFCD01863558
CAS Number: 218301-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR