5-Methyl-4H-1,2,4-triazol-3-ylamine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Methyl-4H-1,2,4-triazol-3-amine hydrochloride
036195-5G
036195-25G
101840-872EA
150.77
USD
101840-872
101840-874
5-Methyl-4H-1,2,4-triazol-3-ylamine hydrochloride ≥95%
5-Methyl-4H-1,2,4-triazol-3-ylamine hydrochloride
Hazard: Irritant
Formula:
C₃H₇ClN₄ MW: 134.57 g/mol |
MDL Number:
MFCD26407488 CAS Number: 23350-30-3 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...