Ethyl-1-(cyanomethyl)piperidine-4-carboxylate
Supplier: Matrix Scientific
036123-1G
036123-5G
101840-726EA
320
USD
101840-726
101840-728
Ethyl-1-(cyanomethyl)piperidine-4-carboxylate
Ethyl-1-(cyanomethyl)piperidine-4-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₆N₂O₂ |
MDL Number:
MFCD10568303 CAS Number: 460094-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...