1H,1H,8H-Perfluoro-1-octanol

Supplier: Matrix Scientific

Synonyms: 1H,1H,8H-Tetradecafluorooctan-1-ol

002861-1G 002861-5G
101784-784EA 161.54 USD
101784-784 101784-786
1H,1H,8H-Perfluoro-1-octanol
1H,1H,8H-Perfluoro-1-octanol

Hazard: Irritant

Formula: C₈H₄F₁₄O
MW: 382.1 g/mol
MDL Number: MFCD01631687
CAS Number: 10331-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR