4-Chloro-6-fluoroquinoline

Supplier: Matrix Scientific

002781-500MG 002781-1G
101784-642EA 144.62 USD
101784-642 101784-644
4-Chloro-6-fluoroquinoline
4-Chloro-6-fluoroquinoline

Hazard: Irritant

Formula: C₉H₅ClFN
MW: 181.60 g/mol
Melting Pt: 76-77°C
MDL Number: MFCD00278783
CAS Number: 391-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR