2-Chloro-6-fluoro-3-methylcinnamic acid

Supplier: Matrix Scientific

002760-1G 002760-5G
101784-604EA 84.62 USD
101784-604 101784-606
2-Chloro-6-fluoro-3-methylcinnamic acid
2-Chloro-6-fluoro-3-methylcinnamic acid

Hazard: Irritant

Formula: C₁₀H₈ClFO₂
MDL Number: MFCD01631396
CAS Number: 261762-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR