2-Chloro-6-fluoro-3-methylbenzyl alcohol

Supplier: Matrix Scientific

002757-1G 002757-5G
101784-600EA 44.62 USD
101784-600 101784-602
2-Chloro-6-fluoro-3-methylbenzyl alcohol
2-Chloro-6-fluoro-3-methylbenzyl alcohol

Hazard: Irritant

Formula: C₈H₈ClFO
MDL Number: MFCD01631362
CAS Number: 261762-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR