2-Chloro-6-fluoro-3-methylbenzamide

Supplier: Matrix Scientific

002754-1G 002754-5G
101784-592EA 46.15 USD
101784-592 101784-594
2-Chloro-6-fluoro-3-methylbenzamide
2-Chloro-6-fluoro-3-methylbenzamide

Hazard: Irritant

Formula: C₈H₇ClFNO
MDL Number: MFCD01631394
CAS Number: 286474-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR