2-Chloro-6-fluoro-3-methylanisole

Supplier: Matrix Scientific

002752-1G 002752-5G
101784-588EA 61.54 USD
101784-588 101784-590
2-Chloro-6-fluoro-3-methylanisole
2-Chloro-6-fluoro-3-methylanisole

Hazard: Irritant

Formula: C₈H₈ClFO
MDL Number: MFCD01631367
CAS Number: 261762-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR