2-Fluoro-6-(trifluoromethyl)phenylacetic acid ≥98%

Supplier: Matrix Scientific

002744-1G 002744-5G
101784-564EA 63.08 USD
101784-564 101784-566
2-Fluoro-6-(trifluoromethyl)phenylacetic acid ≥98%
2-Fluoro-6-(trifluoromethyl)phenylacetic acid

Hazard: Irritant

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00061191
CAS Number: 179946-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR