2-Methyl-5-nitrobenzotrifluoride ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-4-nitro-2-(trifluoromethyl)benzene

002719-1G 002719-5G 002719-25G
101784-520EA 15.38 USD
101784-520 101784-522 101784-524
2-Methyl-5-nitrobenzotrifluoride ≥97%
2-Methyl-5-nitrobenzotrifluoride

Hazard: Toxic

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 34-35°C
MDL Number: MFCD01631684
CAS Number: 89976-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR