2-Fluoro-5-iodobenzotrifluoride ≥97%

Supplier: Matrix Scientific

002703-5G 002703-25G
101784-470EA 26.15 USD
101784-470 101784-472
2-Fluoro-5-iodobenzotrifluoride ≥97%
2-Fluoro-5-iodobenzotrifluoride

Hazard: Irritant/Refrigerate

Formula: C₇H₃F₄I
MDL Number: MFCD00236678
CAS Number: 59382-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR