2-Fluoro-5-(trifluoromethyl)cinnamic acid

Supplier: Matrix Scientific

002643-1G 002643-5G
101784-374EA 33.85 USD
101784-374 101784-376
2-Fluoro-5-(trifluoromethyl)cinnamic acid
2-Fluoro-5-(trifluoromethyl)cinnamic acid

Hazard: Irritant

Formula: C₁₀H₆F₄O₂
MW: 234.15 g/mol
MDL Number: MFCD00236293
CAS Number: 247113-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR