3-Fluoro-5-(trifluoromethyl)phenylacetonitrile ≥98%

Supplier: Matrix Scientific

002639-1G 002639-5G
101784-366EA 30.77 USD
101784-366 101784-368
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile ≥98%
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile

Hazard: Irritant-Harmful

Formula: C₉H₅F₄N
MW: 203.1400 g/mol
MDL Number: MFCD00061181
CAS Number: 239087-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR