(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid
002636-1G
002636-5G
002636-25G
101784-356EA
27.69
USD
101784-356
101784-358
102832-752
(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid ≥98%
(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid
Hazard: Irritant
Formula:
C₉H₆F₄O₂ MW: 222.14 g/mol |
MDL Number:
MFCD00061187 CAS Number: 220227-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...