3-Fluoro-5-(trifluoromethyl)benzophenone ≥97%

Supplier: Matrix Scientific

002627-1G 002627-5G
101784-334EA 61.54 USD
101784-334 101784-336
3-Fluoro-5-(trifluoromethyl)benzophenone ≥97%
3-Fluoro-5-(trifluoromethyl)benzophenone

Hazard: Irritant

Formula: C₁₄H₈F₄O
MDL Number: MFCD00061256
CAS Number: 239087-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR