2-Chloro-5-(trifluoromethyl)benzophenone ≥97%

Supplier: Matrix Scientific

002618-1G 002618-5G
101784-302EA 87.69 USD
101784-302 101784-304
2-Chloro-5-(trifluoromethyl)benzophenone ≥97%
2-Chloro-5-(trifluoromethyl)benzophenone

Hazard: Irritant

Formula: C₁₄H₈ClF₃O
Melting Pt: 39…40 °C
MDL Number: MFCD00039229
CAS Number: 789-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR