(3-Fluoro-4-methylphenyl)acetic acid

Supplier: Matrix Scientific

002583-1G 002583-5G
101784-210EA 47.69 USD
101784-210 101784-212
(3-Fluoro-4-methylphenyl)acetic acid
(3-Fluoro-4-methylphenyl)acetic acid

Hazard: Irritant

Formula: C₉H₉FO₂
MDL Number: MFCD01631536
CAS Number: 261951-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR