3-Fluoro-4-methylcinnamic acid

Supplier: Matrix Scientific

Synonyms: 3-(3-Fluoro-4-methylphenyl)acrylic acid

002580-1G 002580-5G
101784-202EA 75.38 USD
101784-202 101784-204
3-Fluoro-4-methylcinnamic acid
3-Fluoro-4-methylcinnamic acid

Hazard: Irritant

Formula: C₁₀H₉FO₂
MW: 180.18 g/mol
MDL Number: MFCD01631413
CAS Number: 261951-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR