8-Fluoro-4-quinolinol

Supplier: Matrix Scientific

Synonyms: 8-Fluoroquinolin-4-ol, 8-Fluoro-4-hydroxyquinoline

002543-250MG 002543-1G
101784-120EA 30.77 USD
101784-120 101784-122
8-Fluoro-4-quinolinol
8-Fluoro-4-quinolinol

Hazard: Irritant

Formula: C₉H₆FNO
MW: 163.15 g/mol
Melting Pt: 260 °C
MDL Number: MFCD00272395
CAS Number: 63010-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR