3-Chloro-4-fluoro-DL-phenylglycine

Supplier: Matrix Scientific

002489-1G 002489-5G
101783-976EA 106.15 USD
101783-976 101783-978
3-Chloro-4-fluoro-DL-phenylglycine
3-Chloro-4-fluoro-DL-phenylglycine

Hazard: Irritant

Formula: C₈H₇ClFNO₂
MW: 203.6 g/mol
MDL Number: MFCD01631397
CAS Number: 261762-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR