3-Fluoro-4-(trifluoromethyl)cinnamic acid

Supplier: Matrix Scientific

002442-1G 002442-5G
101783-932EA 29.23 USD
101783-932 101783-934
3-Fluoro-4-(trifluoromethyl)cinnamic acid
3-Fluoro-4-(trifluoromethyl)cinnamic acid

Hazard: Irritant

Formula: C₁₀H₆F₄O₂
MDL Number: MFCD00236295
CAS Number: 231291-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR