2-Fluoro-4-(trifluoromethyl)cinnamic acid

Supplier: Matrix Scientific

Synonyms: 3-(2-Fluoro-4-(trifluoromethyl)phenyl)acrylic acid

002441-1G 002441-5G
101783-928EA 232.31 USD
101783-928 101783-930
2-Fluoro-4-(trifluoromethyl)cinnamic acid
2-Fluoro-4-(trifluoromethyl)cinnamic acid

Hazard: Irritant

Formula: C₁₀H₆F₄O₂
MW: 234.1500 g/mol
MDL Number: MFCD00236292
CAS Number: 262608-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR