3-Fluoro-4-(trifluoromethyl)benzophenone ≥97%
Supplier: Matrix Scientific
002436-1G
002436-5G
101783-924EA
47.69
USD
101783-924
101783-926
3-Fluoro-4-(trifluoromethyl)benzophenone ≥97%
3-Fluoro-4-(trifluoromethyl)benzophenone
Hazard: Irritant
Formula:
C₁₄H₈F₄O |
MDL Number:
MFCD00236282 CAS Number: 243128-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...