3-Fluoro-4-(trifluoromethyl)benzamide ≥97%

Supplier: Matrix Scientific

002433-1G 002433-5G
101783-914EA 38.46 USD
101783-914 101783-916
3-Fluoro-4-(trifluoromethyl)benzamide ≥97%
3-Fluoro-4-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₈H₅F₄NO
CAS Number: 247170-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR