2-Fluoro-4-(trifluoromethyl)benzophenone ≥97%

Supplier: Matrix Scientific

002424-1G 002424-5G
101783-898EA 40 USD
101783-898 101783-900
2-Fluoro-4-(trifluoromethyl)benzophenone ≥97%
2-Fluoro-4-(trifluoromethyl)benzophenone

Hazard: Irritant

Formula: C₁₄H₈F₄O
MDL Number: MFCD00061253
CAS Number: 207974-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR