Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane

Supplier: Matrix Scientific

002419-1G 002419-5G
101783-880EA 36.92 USD
101783-880 101783-882
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane

Hazard: Irritant

Formula: C₂₇H₁₆F₆N₂O₆
MW: 578.43 g/mol
Melting Pt: 158…160 °C
MDL Number: MFCD00317556
CAS Number: 69563-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR