3-Chloro-4,5-difluorobenzotrifluoride ≥97%

Supplier: Matrix Scientific

002415-1G 002415-5G
101783-868EA 67.69 USD
101783-868 101783-870
3-Chloro-4,5-difluorobenzotrifluoride ≥97%
3-Chloro-4,5-difluorobenzotrifluoride

Hazard: Irritant

Formula: C₇H₂ClF₅
MDL Number: MFCD00153090
CAS Number: 77227-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR