5-Chloro-3-methylbenzo[b]thiophene-2-sulfonamide

Supplier: Matrix Scientific

002372-1G 002372-5G
101783-784EA 60 USD
101783-784 101783-786
5-Chloro-3-methylbenzo[b]thiophene-2-sulfonamide
5-Chloro-3-methylbenzo[b]thiophene-2-sulfonamide

Hazard: Irritant

Formula: C₉H₈ClNO₂S₂
MW: 261.75 g/mol
MDL Number: MFCD00210142
CAS Number: 190659-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR