2-Methyl-3-(trifluoromethyl)benzamide

Supplier: Matrix Scientific

002313-1G 002313-5G
101783-672EA 100 USD
101783-672 101783-674
2-Methyl-3-(trifluoromethyl)benzamide
2-Methyl-3-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₉H₈F₃NO
MDL Number: MFCD01631495
CAS Number: 251651-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR