4-Fluoro-3-(trifluoromethyl)anisole

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-methoxybenzotrifluoride, 1-Fluoro-4-methoxy-2-(trifluoromethyl)benzene

002312-1G 002312-5G
101783-668EA 49.23 USD
101783-668 101783-670
4-Fluoro-3-(trifluoromethyl)anisole
4-Fluoro-3-(trifluoromethyl)anisole

Hazard: Flammable

Formula: C₈H₆F₄O
MW: 194.13 g/mol
MDL Number: MFCD01631405
CAS Number: 127271-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR