2-Fluoro-3-(trifluoromethyl)anisole

Supplier: Matrix Scientific

002311-1G 002311-5G
101783-664EA 81.54 USD
101783-664 101783-666
2-Fluoro-3-(trifluoromethyl)anisole
2-Fluoro-3-(trifluoromethyl)anisole

Hazard: Irritant

Formula: C₈H₆F₄O
MW: 194.13 g/mol
Flash Pt: 71°C
MDL Number: MFCD01631563
CAS Number: 151868-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR